Learn More
Biomatik Corporation Human Meningioma Expressed Antigen 5 (MGEA5) CLIA Kit
Supplier:  Biomatik Corporation EKU09553
Encompass_Preferred
Human Meningioma Expressed Antigen 5 (MGEA5) CLIA Kit for detecting MGEA5 in urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O60502. Gene ID: 10724. Target Synonyms: NAGase; MEA5; NCOAT; Hexosaminidase C; N-Acetylhexosaminidase; Hyaluronidase; Meningioma-Expressed Antigen 5; Nuclear Cytoplasmic O-GlcNAcase And Acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8498