Learn More
Sino Biological Human MEP1B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10708T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MEP1B / Meprin beta (rh MEP1B / Meprin beta; Catalog#10708-H08H; NP_005916.2; Met1-Thr652). MEP1B / Meprin beta specific IgG was purified by Human MEP1B / Meprin beta affinity chromatography.
Catalog No. 50-237-1406