Learn More
BIOHIPPO Human MEPE Microsample
SDP
Catalog No. 5000315236
Encompass
Human MEPE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MEPE in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 137 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MEPE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MEPE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MEPE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15236 Supplier BIOHIPPO Supplier No. BHE15212186
Human MEPE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MEPE in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 137 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MEPE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MEPE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MEPE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color