Learn More
ACROBiosystems ACROBiosystems Human MERTK / Mer Protein, Fc Tag
Supplier:  ACROBiosystems MEKH52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human MERTK, Fc Tag (MEK-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession # Q12866-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 79.1 Kda. The protein migrates as 100-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant Human (NSO-derived) at 5 μg/mL (100 μL/well) can bind Human MERTK, Fc Tag (Cat. No. MEK-H5253) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-210-6408