Learn More
Sino Biological Anti-MESDC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10949RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MESDC2 (rh MESDC2; Catalog#10949-H08H; NP_055969.1; Ala 34-Lys 230). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5023