Learn More
ACROBiosystems PE-Labeled Human Mesothelin (296-580), His Tag (MSN-HP2H8) is produced via conjugation of PE to Human Mesothelin (296-580), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-
Supplier:  ACROBiosystems MSNHP2H8200TESTS
Encompass_Preferred
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
Catalog No. 50-225-3847