Learn More
ACROBiosystems Alexa Fluor 647-Labeled Human Mesothelin / MSLN (296-580) Protein, His TagStar Staining
Catalog No. 502331791
Encompass_Preferred
Alexa Fluor 647-Labeled Human Mesothelin (296-580), His Tag (MSN-HA2H5) is produced via conjugation of AF647 to Human Mesothelin (296-580), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), His Tag is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1).
Catalog No. 50-233-1791 Supplier ACROBiosystems Supplier No. MSNHA2H525UG
Alexa Fluor 647-Labeled Human Mesothelin (296-580), His Tag (MSN-HA2H5) is produced via conjugation of AF647 to Human Mesothelin (296-580), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), His Tag is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1).