Learn More
ACROBiosystems Biotinylated Human Mesothelin (296-600), His,Avitag (MSN-H82E7) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Thr 600 (Accession # AAH09272.1).
Supplier:  ACROBiosystems MSNH82E725UG
Encompass_Preferred
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
Catalog No. 50-223-1392