Learn More
ACROBiosystems ACROBiosystems Human Mesothelin-coupled Magnetic Beads
Catalog No. 502107058
Encompass_Preferred
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications.
Catalog No. 50-210-7058 Supplier ACROBiosystems Supplier No. MBSK02410MG
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications.