Learn More
Sino Biological Recombinant Human c-MET/HGFR Protein (Site-Specific PE Conjugation, ECD, His Tag)
Catalog No. 502992685
Encompass_Preferred
A DNA sequence encoding the Human MET (NP_000236.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with PE (Excitation = 495-566 nm, Emission Max. = 575 nm).
Catalog No. 50-299-2685 Supplier Sino Biological Supplier No. 10692H86HSP25
A DNA sequence encoding the Human MET (NP_000236.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with PE (Excitation = 495-566 nm, Emission Max. = 575 nm).