Learn More
Sino Biological Anti-c-MET/HGFR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10692MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human c-Met (rh c-Met; Catalog#10692-H08H; NP_000236.2; Met 1-Thr 932). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8244