Learn More
Sino Biological Human MET MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10692R271A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HGFR / c-MET (rh HGFR / c-MET; Catalog#10692-H08H; NP_000236.2; Met 1-Thr 932) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-3136