Learn More
Biomatik Corporation Human Metal Regulatory Transcription Factor 1 (MTF1) ELISA Kit
Supplier:  Biomatik Corporation EKU12393
Encompass_Preferred
Human Metal Regulatory Transcription Factor 1 (MTF1) ELISA Kit for detecting MTF1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14872. Gene ID: 4520. Target Synonyms: ZRF; MRE-binding transcription factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7972