Learn More
Biomatik Corporation Human Metallothionein 1M (MT1M) ELISA Kit
Supplier:  Biomatik Corporation EKU05920
Encompass_Preferred
Human Metallothionein 1M (MT1M) ELISA Kit for detecting MT1M in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200ng/ml. Sensitivity: 0.97ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8N339. Gene ID: 4499. Target Synonyms: MT1K. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6962