Learn More
Sino Biological Human metapneumovirus (HMPV) Fusion glycoprotein Antibody, Rabbit PAb, Antigen Affinity Purified 100 µL
Catalog No. 5000748940
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human metapneumovirus (HMPV) Fusion glycoprotein ( Catalog#40993-V08H1; Q6WB98). Human metapneumovirus (HMPV) Fusion glycoprotein specific IgG was purified by Human metapneumovirus (HMPV) Fusion glycoprotein affinity chromatography.
Catalog No. 50-007-48940 Supplier Sino Biological Supplier No. 40993T62100
Produced in rabbits immunized with purified, recombinant Human metapneumovirus (HMPV) Fusion glycoprotein ( Catalog#40993-V08H1; Q6WB98). Human metapneumovirus (HMPV) Fusion glycoprotein specific IgG was purified by Human metapneumovirus (HMPV) Fusion glycoprotein affinity chromatography.