Learn More
ACROBiosystems Human metapneumovirus Post-fus
Catalog No. 502534216
Encompass_Preferred
Human metapneumovirus Post-fusion glycoprotein F0 His Tag (RSF-V52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 19- Thr 489 (Accession # Q6WB98).
Catalog No. 50-253-4216 Supplier ACROBiosystems Supplier No. RSFV52H5100UG
Human metapneumovirus Post-fusion glycoprotein F0 His Tag (RSF-V52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 19- Thr 489 (Accession # Q6WB98).