Learn More
Sino Biological Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0320/2010/A) glycoprotein G Protein (His Tag)
Supplier:  Sino Biological 40791V08H100
Encompass_Preferred
A DNA sequence encoding the Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0320/2010/A) glycoprotein G (AHV79442.1) (Asn52-Asn228) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-228-2236