Learn More
Sino Biological Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G Antibody, Rabbit PAb, Antigen Affinity Purified 100 µL
Catalog No. 5000749942
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G ( Catalog#40798-V08H; AHV79646.1; Asp52-Ser238). Glycoprotein G specific IgG was purified by Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G affinity chromatography.
Catalog No. 50-007-49942 Supplier Sino Biological Supplier No. 40798T30100
Produced in rabbits immunized with purified, recombinant Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G ( Catalog#40798-V08H; AHV79646.1; Asp52-Ser238). Glycoprotein G specific IgG was purified by Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G affinity chromatography.