Learn More
Sino Biological Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G Protein (His Tag) 100ug
Supplier:  Sino Biological 40798V08H100
Encompass_Preferred
A DNA sequence encoding the Human metapneumovirus (strain HMPV/Homo sapiens/PER/CFI0466/2010/B) glycoprotein G (AHV79646.1) (Asp52-Ser238) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6887