Learn More
Biomatik Corporation Human Methionine Sulfoxide Reductase A (MSRA) ELISA Kit
Supplier:  Biomatik Corporation EKU05935
Encompass_Preferred
Human Methionine Sulfoxide Reductase A (MSRA) ELISA Kit for detecting MSRA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UJ68. Gene ID: 4482. Target Synonyms: PMSR; Peptide Methionine Sulfoxide Reductase; Protein-methionine-S-oxide reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4390