Learn More
Zymo Research Corporation Human Methylated & Non-Methylated DNA Set (DNA w/ primers)
SDP
Catalog No. 50444309
Encompass_Preferred
The Human HCT116 DKO Non-methylated DNA is purified from cells that contain genetic knockouts of both DNA methyltransferasesDNMT1 (-/-) and DNMT3b (-/-). The DNA derived from HCT116 DKO cells has a low level of DNA methylation and can be used as a control for DNA methylation analysis. The Human HCT116 DKO Methylated DNA is purified HCT116 DKO DNA that has been enzymatically methylated at all cytosine positions comprising CG dinucleotides by M.SssI methyltransferase2 (EC 2.1.1.37) and can be used as a positive control for DNA methylation analysis.
Catalog No. 50-444-309 Supplier Zymo Research Corporation Supplier No. D5014
The Human HCT116 DKO Non-methylated DNA is purified from cells that contain genetic knockouts of both DNA methyltransferasesDNMT1 (-/-) and DNMT3b (-/-). The DNA derived from HCT116 DKO cells has a low level of DNA methylation and can be used as a control for DNA methylation analysis. The Human HCT116 DKO Methylated DNA is purified HCT116 DKO DNA that has been enzymatically methylated at all cytosine positions comprising CG dinucleotides by M.SssI methyltransferase2 (EC 2.1.1.37) and can be used as a positive control for DNA methylation analysis.