Learn More
Zymo Research Corporation Human Methylated & Non-Methylated (WGA) DNA Set (DNA w/ primers)
SDP
Catalog No. 501251525
Encompass_Preferred
The Human WGA Methylated & Non-methylated DNA Set isgenerated using phi29 DNA polymerase based whole genome amplification techniques from HCT116 DKO cell line derived genomic DNA (Human HCT116 DKO Non-methylated DNA). The Human WGA Methylated DNA is Human WGA Non-methylated DNA that has been enzymatically methylated at all double-stranded CGdinucleotides using M.SssI methyltransferase and can be used as a positive control for DNA methylation analysis.
Catalog No. 50-125-1525 Supplier Zymo Research Corporation Supplier No. D5013
The Human WGA Methylated & Non-methylated DNA Set isgenerated using phi29 DNA polymerase based whole genome amplification techniques from HCT116 DKO cell line derived genomic DNA (Human HCT116 DKO Non-methylated DNA). The Human WGA Methylated DNA is Human WGA Non-methylated DNA that has been enzymatically methylated at all double-stranded CGdinucleotides using M.SssI methyltransferase and can be used as a positive control for DNA methylation analysis.