Learn More
Biomatik Corporation Human Methyltransferase Like Protein 16 (METTL16) ELISA Kit
Supplier:  Biomatik Corporation EKU12101
Encompass_Preferred
Human Methyltransferase Like Protein 16 (METTL16) ELISA Kit for detecting METTL16 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86W50. Gene ID: 79066. Target Synonyms: METT10D; Methyltransferase 10 Domain Containing Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8029