Learn More
Sino Biological Anti-METTL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11525MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human METTL1 (rh METTL1; Catalog#11525-H07E; NP_005362.3; Asp 32-Gln 265). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1897