Learn More
Sino Biological Anti-METTL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11525RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human METTL1 (rh METTL1; Catalog#11525-H07E; NP_005362.3; Asp32-Gln265). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8369