Learn More
Bon Opus Biosciences Recombinant Human MFI2/MTf (N-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP102850UG
Encompass_Preferred
Recombinant Human MFI2/MTf (N-hFc) is produced by Human Cells. The target gene encoding Gly20-Cys709 is expressed with a N-hFc tag. Accession# P08582. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1028
Catalog No. 50-217-2627