Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MFSD1 Synthetic Peptide

Catalog No. PEP1162 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1162 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1162 Supplier Invitrogen™ Supplier No. PEP1162
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 18 amino acids near the center of human MFSD1.

MFSD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 64747
Species Human
Purification Method purified
Accession Number Q9H3U5
Common Name MFSD1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1200003O06Rik; hypothetical protein LOC641486; major facilitator superfamily domain containing 1; major facilitator superfamily domain-containing protein 1; Major facilitator superfamily domain-containing protein 1-like protein; Mfsd1; mfsd1a; RGD1305751; SMAP4; SMAP-4; Smooth muscle cell-associated protein 4; UG0581B09; zgc:123187
Gene Symbol MFSD1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.