Learn More
BIOHIPPO Human MHCDRb1 Microsample
SDP
Catalog No. 5000314517
Encompass
Human MHCDRb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCDRb1 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 38 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MHCDRb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MHCDRb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MHCDRb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14517 Supplier BIOHIPPO Supplier No. BHE15211445
Human MHCDRb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCDRb1 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 38 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MHCDRb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MHCDRb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MHCDRb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color