Learn More
BIOHIPPO Human MHCF Microsample
SDP
Catalog No. 5000313877
Encompass
Human MHCF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCF in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MHCF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MHCF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MHCF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13877 Supplier BIOHIPPO Supplier No. BHE15210581
Human MHCF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MHCF in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MHCF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MHCF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MHCF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color