Learn More
BIOHIPPO Human MIA1 Microsample
SDP
Catalog No. 5000315528
Encompass
Human MIA1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIA1 in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MIA1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MIA1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MIA1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15528 Supplier BIOHIPPO Supplier No. BHE15212562
Human MIA1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIA1 in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MIA1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MIA1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MIA1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color