Learn More
ACROBiosystems Human MICA 25ug
Catalog No. 502534435
Encompass_Preferred
Biotinylated Human MICA Protein FcAvitag (MIA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # Q29983-1).
Catalog No. 50-253-4435 Supplier ACROBiosystems Supplier No. MIAH82F625UG
Biotinylated Human MICA Protein FcAvitag (MIA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # Q29983-1).