Learn More
BIOHIPPO Human MICA Microsample
SDP
Catalog No. 5000315515
Encompass
Human MICA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MICA in serum plasma tissue homogenates cell lysates urine cerebrospinal fluid cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 36 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MICA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MICA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MICA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15515 Supplier BIOHIPPO Supplier No. BHE15210688
Human MICA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MICA in serum plasma tissue homogenates cell lysates urine cerebrospinal fluid cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 36 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MICA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MICA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MICA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color