Learn More
Sino Biological Anti-MICB Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10759MM12F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MICB / MIC-B (rh MICB; Catalog#10759-H08H; NP_005922.2; Met 1-Gly 298) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1808