Learn More
Sino Biological Recombinant Anti-MICB Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10759R102100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MICB (rh MICB ; Catalog#10759-H08H; NP_005922.2; Met1-Gly298).
Catalog No. 50-241-1729