Learn More
Sino Biological Anti-MICB Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10759MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MICB (rh MICB ; Catalog#10759-H08H; NP_005922.2; Met1-Gly298). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2118