Learn More
Sino Biological Human MICB PAb 100ul (Sino Biological)
Supplier:  Sino Biological 90079R001100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MICB (rh MICB; Catalog#10759-H08H; NP_005922.2; Met1-Gly298). MICB specific IgG was purified by Human MICB affinity chromatography.
Catalog No. 50-237-4547