Learn More
ACROBiosystems Human MICB Protein His Tag
Supplier:  ACROBiosystems MIBH52H3100UG
Encompass_Preferred
Human MICB Protein His Tag (MIB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Thr 308 (Accession # NP_005922.2).
Catalog No. 50-253-4432