Learn More
Sciencell Research Laboratories Human Microglia
Catalog No. NC1614510
Encompass
HM from ScienCell Research Laboratories are isolated from human brain. HM are cryopreserved after purification and delivered frozen. Each vial contains >1 x 106 cells in 1 ml volume.
Catalog No. NC1614510 Supplier Sciencell Research Laboratories Supplier No. 1900
HM from ScienCell Research Laboratories are isolated from human brain. HM are cryopreserved after purification and delivered frozen. Each vial contains >1 x 106 cells in 1 ml volume.