Learn More
Biomatik Corporation Human Microseminoprotein Beta (MSMb) ELISA Kit
Supplier:  Biomatik Corporation EKU05945
Encompass_Preferred
Human Microseminoprotein Beta (MSMb) ELISA Kit for detecting MSMb in serum, plasma, seminal plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08118. Gene ID: 4477. Target Synonyms: MSM-B; IGBF; MSP; MSPB; PN44; PRPS; PSP; PSP57; PSP94; Immunoglobulin-binding factor; Prostate secreted seminal plasma protein; plasma beta-inhibin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7771