Learn More
Biomatik Corporation Human Microtubule Associated Protein 4 (MAP4) ELISA Kit
Supplier:  Biomatik Corporation EKU05952
Encompass_Preferred
Human Microtubule Associated Protein 4 (MAP4) ELISA Kit for detecting MAP4 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.101ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27816. Gene ID: 4134. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8871