Learn More
Biomatik Corporation Human Microtubule Associated Serine/Threonine Kinase 2 (MAST2) ELISA Kit
Supplier:  Biomatik Corporation EKU05958
Encompass_Preferred
Human Microtubule Associated Serine/Threonine Kinase 2 (MAST2) ELISA Kit for detecting MAST2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P0Q8. Gene ID: 23139. Target Synonyms: MAST205; MTSSK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6515