Learn More
ACROBiosystems Human MIF 25ug
Catalog No. 502534430
Encompass_Preferred
Biotinylated Human MIF HisAvitag (MIF-H82E7) is expressed from human 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession # P14174-1).
Catalog No. 50-253-4430 Supplier ACROBiosystems Supplier No. MIFH82E725UG
Biotinylated Human MIF HisAvitag (MIF-H82E7) is expressed from human 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession # P14174-1).