Learn More
Sino Biological Anti-MIF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10246T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MIF (rh MIF; Catalog#10246-H08H; P14174; Pro2-Ala115). MIF specific IgG was purified by Human MIF affinity chromatography.
Catalog No. 50-240-9110