Learn More
Biomatik Corporation Human Milk Fat Globule EGF Factor 8 (MFGE8) CLIA Kit
Supplier:  Biomatik Corporation EKU11953
Encompass_Preferred
Human Milk Fat Globule EGF Factor 8 (MFGE8) CLIA Kit for detecting MFGE8 in serum, plasma, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q08431. Gene ID: 4240. Target Synonyms: HMFG; MFGM; SED1; BA46; EDIL1; OAcGD3S; SPAG10; hP47; Medin; Lactadherin; Sperm Associated Antigen 10; Sperm Surface Protein hP47; Milk fat globule-EGF factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5431