Learn More
Medchemexpress LLC Human milk lysozyme | 12671-19-1 | 96% | 250 MG
SDP
Catalog No. 5000396576
Encompass_Preferred
Human milk lysozyme is a key defense factor found in human milk, playing a crucial role in protecting the gastrointestinal tract of newborns against bacterial infection. In vitro studies demonstrate its superior activity against skin pathogens such as Pr. acnes and C. minutissimum. While it shows no activity against Gram-negative bacteria like E. coli, Salm. typhimurium, and Bord. bronchiseptica, it exhibits weak activity against Kleb. pneumonieae and Ps. aeruginosa. It also lacks activity against Staph. epidermidis and shows weak activity against Staph. aureus and Str. zooepidicus. This product is intended for research use only.
  • Key defense factor in newborn gastrointestinal protection
  • Exhibits superior activity against Pr. acnes and C. minutissimum
  • Shows weak activity against Kleb. pneumonieae and Ps. aeruginosa
  • Intended for research use only
Catalog No. 50-003-96576 Supplier Medchemexpress LLC Supplier No. 250MGHYP3021
Human milk lysozyme is a key defense factor found in human milk, playing a crucial role in protecting the gastrointestinal tract of newborns against bacterial infection. In vitro studies demonstrate its superior activity against skin pathogens such as Pr. acnes and C. minutissimum. While it shows no activity against Gram-negative bacteria like E. coli, Salm. typhimurium, and Bord. bronchiseptica, it exhibits weak activity against Kleb. pneumonieae and Ps. aeruginosa. It also lacks activity against Staph. epidermidis and shows weak activity against Staph. aureus and Str. zooepidicus. This product is intended for research use only.
  • Key defense factor in newborn gastrointestinal protection
  • Exhibits superior activity against Pr. acnes and C. minutissimum
  • Shows weak activity against Kleb. pneumonieae and Ps. aeruginosa
  • Intended for research use only