Learn More
Sino Biological Human MILR1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 16083H02H1
Encompass_Preferred
A DNA sequence encoding the human MILR1 (NP_001078892.1) (Met1-Lys227) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3082