Learn More
Biomatik Corporation Human Mineralocorticoid Receptor (MR) ELISA Kit
Supplier:  Biomatik Corporation EKU05966
Encompass_Preferred
Human Mineralocorticoid Receptor (MR) ELISA Kit for detecting MR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08235. Gene ID: 4306. Target Synonyms: NR3C2; MLR; MCR; Aldosterone Receptor; Nuclear Receptor Subfamily 3 Group C Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6053