Learn More
Biomatik Corporation Human Minichromosome Maintenance Deficient 5 (MCM5) ELISA Kit
Supplier:  Biomatik Corporation EKU05969
Encompass_Preferred
Human Minichromosome Maintenance Deficient 5 (MCM5) ELISA Kit for detecting MCM5 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33992. Gene ID: 4174. Target Synonyms: CDC46; P1-CDC46; Cell Division Cycle 46; DNA replication licensing factor MCM5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8868