Learn More
FUJIFILM BIOSCIENCES INC Human MIP-1 alpha (CCL3), Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100155UG
Encompass_Preferred
Macrophage inflammatory protein-1 alpha (MIP-1 A), also known as CCL3, is a cytokine produced by macrophages.  MIP-1 A binds the chemokine receptors CCR1, CCR4 and CCR5 to induce inflammatory responses, including the recruitment of granulocytes and neutrophil superoxide production.  The MIP-1 A and MIP-1 B heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 
Catalog No. 50-990-696