Learn More
FUJIFILM BIOSCIENCES INC Human MIP-1b (CCL4), Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10013410UG
Encompass_Preferred
Macrophage inflammatory protein-1 beta (MIP-1 B), also known as CCL4, is  produced by macrophages and functions as a mitogen-inducible cytokine.  MIP-1 B signals through the chemokine receptor CCR5 to chemoattract immune cells.  MIP-1 B induces inflammatory responses, including neutrophil superoxide production.  The MIP-1 B and MIP-1 A heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 
Catalog No. 50-900-490